Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cullin 4a Antibody - N-terminal, Novus Biologicals™
SDP

Catalog No. NBP178040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178040 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178040 Supplier Novus Biologicals Supplier No. NBP178040
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cullin 4a Polyclonal specifically detects Cullin 4a in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Cullin 4a
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 85 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation Antiserum with 0.01% Sodium Azide
Gene Accession No. AAR13072
Gene Alias CUL-4A, cullin 4A, cullin-4A
Gene Symbols CUL4A
Host Species Rabbit
Immunogen This Cullin 4a Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the N-Terminal region near amino acids 1-25 of Human Cullin 4a coupled to KLH. (Uniprot: Q13619)
Molecular Weight of Antigen 87.68 kDa
Purification Method Delipidation and Defibrination
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8451
Test Specificity This product reacts with human Cullin 4A. Cross reactivity is expected against mouse Cul4A based on a high degree of sequence identity.
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.