Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cullin-7 Antibody - C-terminus, Novus Biologicals™
SDP

Catalog No. NBP178042 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178042 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178042 Supplier Novus Biologicals Supplier No. NBP178042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Cullin-7 Polyclonal specifically detects Cullin-7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Cullin-7
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 85 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500
Formulation Antiserum with 0.01% Sodium Azide
Gene Accession No. NP_055595
Gene Alias CUL-7, cullin 7, cullin-7, KIAA0076dJ20C7.5
Gene Symbols CUL7
Host Species Rabbit
Immunogen This Cullin-7 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the C-Terminus region near amino acids 1675-1698 of Human Cullin-7 coupled to KLH. (Uniprot: Q14999)
Molecular Weight of Antigen 191.16 kDa
Purification Method Delipidation and Defibrination
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 9820
Test Specificity This product reacts with human and mouse Cullin 7. Cross reactivity with other human cullins may occur.
Target Species Human, Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.