Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cullin-7 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB291685 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291685 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291685 Supplier Novus Biologicals Supplier No. NB100602F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cullin-7 Polyclonal specifically detects Cullin-7 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen Cullin-7
Applications Immunoprecipitation, Western Blot (Negative)
Classification Polyclonal
Conjugate FITC
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias CUL-7, cullin 7, cullin-7, KIAA0076dJ20C7.5
Host Species Rabbit
Immunogen A synthetic peptide, mapping to a region between residue 1650 and the C-terminus (residue 1698) of human Cullin homolog 7 using the numbering given in SwissProt entry Q14999 (GeneID 9820).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 9820
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.