Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cumene Hydroperoxide, 77%, Spectrum™ Chemical
SDP

Catalog No. 18601894 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
200 L
Packaging:
Amber Glass Bottle

Catalog No. 18-601-894

Supplier: Spectrum Chemical Manufacturing Corporation C2313200LTBL

Only null left
Add to Cart
Add to Cart
Request a Quote

C2313, 80-15-9, C9H12O2

Spectrum™ Chemical Cumene Hydroperoxide , also known as Cumyl Hydroperoxide, is used as an oxidizing agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 80-15-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002129
InChI Key YQHLDYVWEZKEOX-UHFFFAOYSA-N
IUPAC Name 2-phenylpropane-2-peroxol
SMILES CC(C)(OO)C1=CC=CC=C1

Specifications

CAS Max % 100%
Boiling Point 53°C
Packaging Amber Glass Bottle
Quantity 200 L
UN Number UN3109
Percent Purity 77%
Grade Reagent
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.