Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Globe Scientific Cuvettes for Ciba Corning 550 Express
Click to view available options
Includes:
24 disposable bags
Cuvettes only
Description
- For use with the Bayer 550 Express & Express Plus analyzers
- Cuvettes are packaged in cartridges for easy loading into the analyzers
- Each cartridge contains 13 (5-place) cuvette segments
- Polymethylmethacrylate (PMMA)
- Natural color
- Available with 24 disposable bags (21-200-117)
Specifications
Specifications
| Color | Natural |
| Material | PMMA |
| Includes | Cuvettes only |
| No. per Case | 200 |
| Quantity | 200/Cs. |
| For Use With (Equipment) | Bayer 550 Express & Express Plus Analyzers |
| Product Type | Cuvette |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction