Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUZD1 Antibody (M4P6G6), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309801 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309801 Supplier Novus Biologicals Supplier No. NBP250186MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CUZD1 Monoclonal antibody specifically detects CUZD1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen CUZD1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone M4P6G6
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias CUB and zona pellucida-like domain-containing protein 1, CUB and zona pellucida-like domains 1, CUB and ZP domain-containing protein 1, ERG-1, estrogen regulated gene 1, Transmembrane protein UO-44, UO-44
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide - RHFVNQRADY
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 50624
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.