Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CVT 10216

Click to view available options
Quantity:
10 mg
50 mg
Description
CVT 10216 is a potent and selective, reversible inhibitor of aldehyde dehydrogenase 2 (ALDH2) (IC50 values are 29 and 1300 nM for ALDH2 and ALDH1, respectively). Exhibits anxiolytic effects in rat models.
Specifications
Specifications
Quantity | 50 mg |
Product Type | CVT 10216 |
Purity | >98% |
Molecular Weight (g/mol) | 465.48 |
Inhibitors | CVT 10216 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction