Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CW 008

Click to view available options
Quantity:
10 mg
50 mg
Description
CW 008 is a cAMP/PKA/CREB pathway activator. Promotes osteogenic differentiation of human bone marrow-derived MSCs in vitro. Increases bone mass and bone volume density in ovariectomized mice.
Specifications
Specifications
Chemical Name or Material | 4-Fluoro-N-[5-fluoro-6-(5-methoxypyrazolo[1,5-a]pyridin-3-yl)-1H-pyrazolo[3,4-b]pyridin-3-yl]benzamide |
CAS | 1134613-19-6 |
Quantity | 10 mg |
Target | Protein Kinase A Activators |
Molecular Formula | C21H14F2N6O2 |
Formula Weight | 420.37 |
Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction