Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CX 546
GREENER_CHOICESDP

Catalog No. 298010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-801-0 10 mg
29-805-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-801-0 Supplier Tocris Bioscience™ Supplier No. 2980/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

AMPA receptor potentiator

AMPA receptor potentiator. Binds specifically to the agonist bound non-desensitized receptor, most likely destabilizing the desensitized receptor conformation. Enhances cognitive function in rats. Neuroprotective in mice undergoing repeat ketamine anesthesia.

Chemical Identifiers

CAS 215923-54-9
Molecular Formula C14H17NO3

Specifications

Quantity 10 mg
Formula Weight 247.29
Purity >99%
Molecular Weight (g/mol) 247.294
Chemical Name or Material CX 546
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.