Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CX 614
GREENER_CHOICE

Catalog No. p-5546151
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

AMPAkine; positive allosteric modulator of AMPA

CX 614 is an AMPAkine; positive allosteric modulator of AMPA. Prototypical benzamide which occupies subsite A only. Modulates channel deactivation and desensitization.

Specifications

Quantity 10 mg
Product Type CX 614
Purity >99%
Molecular Weight (g/mol) 247.25
Inhibitors CX 614

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.