Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CX08005

Click to view available options
Quantity:
5 mg
25 mg
Description
CX08005 is a competitive PTP1B inhibitor (IC50 = 0.781 μM). Binds catalytic P-loop of enzyme. Improves insulin sensitivity in obese mice in the glucose tolerance test and the homeostasis model assessment of insulin resistance index (HOMA-IR) . Enhances insulin-induced glucose uptake in adipocytes and myotubes in vitro. Exhibits some selectivity for PTP1B over other protein tyrosine phosphatases, except TCPTP.
Specifications
Specifications
Chemical Name or Material | 2-[[[2-(Tetradecyl)phenyl]amino]carbonyl]benzoic acid |
CAS | 1256341-22-6 |
Quantity | 25 mg |
Molecular Formula | C28H39N04 |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction