Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CX3CL1/Fractalkine Antibody (81506R), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB371853 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371853 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371853 Supplier Novus Biologicals Supplier No. FAB3652RJ
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CX3CL1/Fractalkine Monoclonal specifically detects CX3CL1/Fractalkine in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Neutralization.

Specifications

Antigen CX3CL1/Fractalkine
Applications CyTOF, Flow Cytometry (Intracellular Staining), Neutralization
Classification Monoclonal
Clone 81506R
Conjugate Janelia Fluor 646
Dilution Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1
Host Species Mouse
Immunogen NS0-derived and E. coli-derived Recombinant human CX3CL1/Fracktalkine, Gln25-Arg339, Accession # P78423
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 6376
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.