Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CX3CL1 Mouse anti-Human, Clone: 2F5D5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | CX3CL1 |
|---|---|
| Clone | 2F5D5 |
| Concentration | 0.7 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6426
![]()
|
Proteintech Group Inc
603391IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene belongs to the CX3C subgroup of chemokines, characterized by the number of amino acids located between the conserved cysteine residues. This is the only member of the CX3C subgroup, which contains three amino acids between cysteine residues, resulting in a Cys-X-X-X-Cys configuration. The encoded protein contains an extended mucin-like stalk with a chemokine domain on top, and exists in both a membrane-anchored form where it acts as a binding molecule, or, in soluble form, as a chemotactic cytokine. The mature form of this protein can be cleaved at the cell surface, yielding different soluble forms that can interact with the G-protein coupled receptor, C-X3-C motif chemokine receptor 1 gene product. This gene plays a role in a wide range of diseases, including cancer, vasculitis, neuropathies, atherosclerosis, inflammatory diseases, and in human immunodeficiency virus infections.Specifications
| CX3CL1 | |
| 0.7 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ABCD 3, C X3 C motif chemokine 1, C3Xkine, CX3CL1, CXC3, CXC3C, FKN, fractalkine, neurotactin, NTN, NTT, SCYD1, Small inducible cytokine D1 | |
| CX3CL1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2F5D5 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P78423 | |
| 6376 | |
| CX3CL1 Fusion Protein Ag0161 | |
| Primary | |
| -20°C | |
| CX3CL1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title