Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CX3CR1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB357178 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357178 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357178 Supplier Novus Biologicals Supplier No. NBP176872MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CX3CR1 Polyclonal antibody specifically detects CX3CR1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CX3CR1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKBRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28C-X3-C CKR-1
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to amino acids in a extracellular loop of human CX3CR1. The immunogen is located within amino acids 160 - 210 of CX3CR1. Amino Acid Squence: CLGDYPEVLQEIWP
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, GPCR, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 1524
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.