Learn More
CX3CR1 Rat anti-Human, PE, Clone: 2A9-1, BD
Rat Monoclonal Antibody
$154.00 - $355.50
Specifications
Antigen | CX3CR1 |
---|---|
Clone | 2A9-1 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
The 2A9-1 monoclonal antibody specifically binds to human CX3CR1, which is also known as chemokine (C-C) receptor-like 1 (CCRL1), Beta chemokine receptor-like 1 (CMK-BRL-1), G protein-coupled receptor 13 (GPR13), or GPRV28 (V28). CX3CR1 is a seven transmembrane G protein coupled receptor that is expressed by NK cells, T cells, and monocytes. The cellular expression of CX3CR1 is correlated with high levels of intracellular perforin and granzyme B. CX3CR1 serves as a receptor for fractalkine (CX3CL1). Fractalkine is a transmembrane chemokine of the CX3C family that is expressed on activated endothelial cells, neurons, and astrocytes. Interaction of CX3CR1 with fractalkine initiates cellular adhesive and chemotactic responses.
Specifications
CX3CR1 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.2% sodium azide. | |
IgG2b κ | |
Affinity Purified | |
CCRL1; CMKBRL1; CMKDR1; GPR13; GPRV28; V28; Fractalkine Receptor |
2A9-1 | |
Monoclonal | |
Rat | |
Human | |
Human CX3CR1 Recombinant Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.