Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXADR Antibody (271), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB099614 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB099614 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB099614 Supplier Novus Biologicals Supplier No. NBP289645JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CXADR Monoclonal specifically detects CXADR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Sandwich ELISA Detection.

Specifications

Antigen CXADR
Applications Flow Cytometry, Immunofluorescence, ELISA
Classification Monoclonal
Clone 271
Conjugate Janelia Fluor 646
Formulation 50 mM Sodium Borate
Gene Alias CAR10Coxsackievirus B-adenovirus receptor, CAR4/6, coxsackie virus and adenovirus receptor, coxsackie virus B receptor, coxsackievirus and adenovirus receptor, CVB3 binding protein, CVB3-binding protein, hCAR, HCVADR
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CXADR (NP_001329.1; Met1-Gly237).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Stem Cell Markers, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 1525
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.