Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL1/2/3/GRO Antibody (29702), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB304290 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304290 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304290 Supplier Novus Biologicals Supplier No. FAB2761S

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

CXCL1/2/3/GRO Monoclonal antibody specifically detects CXCL1/2/3/GRO in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen CXCL1/2/3/GRO
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 29702
Conjugate Alexa Fluor 750
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL3, Ala35-Asn107, Accession # P19876
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2919
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.