Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL1/GRO alpha/KC/CINC-1 Antibody (1174A), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB347786 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347786 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347786 Supplier Novus Biologicals Supplier No. FAB4532MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CXCL1/GRO alpha/KC/CINC-1 Monoclonal antibody specifically detects CXCL1/GRO alpha/KC/CINC-1 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen CXCL1/GRO alpha/KC/CINC-1
Applications Flow Cytometry
Classification Monoclonal
Clone 1174A
Conjugate mFluor Violet 450 SE
Dilution Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1
Host Species Rabbit
Immunogen E. coli-derived recombinant mouse CXCL1/GRO alpha/KC/CINC-1, Asn29-Lys96, Accession # P12850
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Membrane Trafficking and Chaperones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2919
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.