Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL1/GRO alpha/KC/CINC-1 Antibody (20326), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB364273 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364273 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364273 Supplier Novus Biologicals Supplier No. FAB275X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCL1/GRO alpha/KC/CINC-1 Monoclonal antibody specifically detects CXCL1/GRO alpha/KC/CINC-1 in Human, Primate samples. It is validated for Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen CXCL1/GRO alpha/KC/CINC-1
Applications Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 20326
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL1/GRO alpha/KC/CINC-1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Membrane Trafficking and Chaperones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2919
Target Species Human, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.