Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL10/IP-10/CRG-2 Antibody (5C7), Novus Biologicals™
SDP

Catalog No. NB297701 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297701 100 μL
NB297700 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297701 Supplier Novus Biologicals Supplier No. NBP326638100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CXCL10/IP-10/CRG-2 Monoclonal antibody specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen CXCL10/IP-10/CRG-2
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 5C7
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias C7, chemokine (C-X-C motif) ligand 10, crg-2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10
Host Species Rabbit
Immunogen A synthesized peptide derived from Human CXCL10/IP-10/CRG-2 [UniProt P02778]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3627
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.