Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL10/IP-10/CRG-2 Antibody (5C7), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CXCL10/IP-10/CRG-2 |
---|---|
Clone | 5C7 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
CXCL10/IP-10/CRG-2 Monoclonal antibody specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
CXCL10/IP-10/CRG-2 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
C7, chemokine (C-X-C motif) ligand 10, crg-2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 | |
A synthesized peptide derived from Human CXCL10/IP-10/CRG-2 [UniProt P02778] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
5C7 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3627 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title