Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL10/IP-10/CRG-2 Antibody (6D4), PerCP, Novus Biologicals™
SDP

Catalog No. NB162725 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162725 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162725 Supplier Novus Biologicals Supplier No. NBP259749PCP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL10/IP-10/CRG-2 Monoclonal antibody specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CXCL10/IP-10/CRG-2
Applications Flow Cytometry
Classification Monoclonal
Clone 6D4
Conjugate PerCP
Formulation PBS
Gene Alias C7, chemokine (C-X-C motif) ligand 10, crg-2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10
Host Species Mouse
Immunogen Recombinant human CXCL10/INP10
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3627
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.