Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL10 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991318
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1318 100 μg
50-199-1319 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1318 Supplier Rockland Immunochemicals Supplier No. 201401B84
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for bovine CXCL10 protein. A BLAST analysis was used to suggest cross-reactivity with CXCL10 from bovine based on 100% homology with the immunizing sequence. Partial reactivity is expected against sheep and goat based on 94-95% homology. Cross-reactivity with CXCL10 from other sources has not been determined.

IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.

Specifications

Antigen CXCL10
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene CXCL10
Gene Accession No. Q2KIQ8
Gene Alias 10 kDa interferon gamma-induced protein; BOS_6808; C Cmotif chemokine; C X C motif chemokine; C7; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 10; chemokine C-X-C motif ligand 10; chemokine CXC-like protein; Crg2; crg-2; CXC; CXC motif chemokine; C-X-C motif chemokine 10; C-X-C motif chemokine 10-like protein; C-X-C motif chemokine ligand 10; CXCL; Cxcl10; CXCL10(1-73); gamma interferon inducible protein 10; gamma IP10; Gamma-IP10; gIP-10; H-IP-10; Ifi10; Inp10; interferon activated gene 10; Interferon-gamma induced protein CRG-2; interferon-gamma-induced protein CRG-2; interferon-gamma-inducible protein 10; interferon-gamma-inducible protein-10; interferon-inducible cytokine IP-10; Interferon-inducible protein 10; IP10; IP-10; M-IP-10; Mob1; mob-1; protein 10 from interferon (gamma)-induced cell line; Protein Mob-1; SCYB10; small inducible cytokine B subfamily (Cys-X-Cys) member 10; small inducible cytokine B subfamily (Cys-X-Cys), member 10; small inducible c
Gene Symbols CXCL10
Host Species Rabbit
Immunogen This protein-A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant protein raised in yeast, corresponding to the 83 amino acids of the mature bovine CXCL10 protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100860873, 442997, 615107
Target Species Bovine, Goat, Ovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.