Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL11/I-TAC Antibody (10C6), Biotin, Novus Biologicals™
SDP

Catalog No. NBP222533B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222533B 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222533B Supplier Novus Biologicals Supplier No. NBP222533B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCL11/I-TAC Monoclonal specifically detects CXCL11/I-TAC in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CXCL11/I-TAC
Applications Western Blot, ELISA
Classification Monoclonal
Clone 10C6
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias Beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TACMGC102770, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11
Gene Symbols CXCL11
Host Species Mouse
Immunogen Full length recombinant protein expressed in E-Coli
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 6373
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.