Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL11/I-TAC Mouse anti-Human, Clone: B-S40, Novus Biologicals™
SDP

Catalog No. NB160254 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160254 500 μg
NB160253 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB160254 Supplier Novus Biologicals Supplier No. NBP318117500UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL11/I-TAC Monoclonal antibody specifically detects CXCL11/I-TAC in Human samples. It is validated for ELISA

Specifications

Antigen CXCL11/I-TAC
Applications ELISA
Classification Monoclonal
Clone B-S40
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias Beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TACMGC102770, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11
Host Species Mouse
Immunogen Recombinant human CXCL11/I-TAC. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells
Purification Method Affinity purified
Quantity 500 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 6373
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.