Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL11/I-TAC Mouse anti-Human, Clone: B-S40, Novus Biologicals™

Mouse Monoclonal Antibody
$366.50 - $699.00
Specifications
Antigen | CXCL11/I-TAC |
---|---|
Clone | B-S40 |
Concentration | 1 mg/ml |
Dilution | ELISA |
Applications | ELISA |
Description
CXCL11/I-TAC Monoclonal antibody specifically detects CXCL11/I-TAC in Human samples. It is validated for ELISASpecifications
CXCL11/I-TAC | |
1 mg/ml | |
ELISA | |
Unconjugated | |
Mouse | |
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research | |
PBS. Sterile-filtered through 0.22 um. | |
6373 | |
IgG1 κ | |
Affinity purified |
B-S40 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TACMGC102770, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11 | |
Recombinant human CXCL11/I-TAC. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
Primary | |
Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title