Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL12/SDF-1 alpha Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP229480 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229480 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229480 Supplier Novus Biologicals Supplier No. NBP229480
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

CXCL12/SDF-1 alpha Polyclonal specifically detects CXCL12/SDF-1 alpha in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Functional.

Specifications

Antigen CXCL12/SDF-1 alpha
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Functional Assay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1-0.2ug/ml, ELISA 0.5-2.0ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 0.5ug/ml, Functional 2.0-4.0ug/ml
Formulation PBS with No Preservative
Gene Alias CXCL12 chemokine (C-X-C motif) ligand 12, IRH, PBSF, SCYB12, SDF1, SDF1A, SDF1B, TLSF, TPAR1
Gene Symbols CXCL12
Host Species Rabbit
Immunogen Antisera to human SDF 1 alpha were raised by repeated immunisations of rabbits with highly purified antigen. Purified IgG prepared by affinity chromatography.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6387
Test Specificity NBP2-29480 recognises human SDF 1 alpha, otherwise known as CXCL12a, a 68 amino acid stromal cell derived CXC chemokine, which arises from alternative splicing of the SDF 1 gene. The two isoforms, SDF 1 alpha and SDF 1 beta, share an identical amino acid sequence, except for an additional four residues in the C terminal region of the beta isoform.
Reconstitution Reconstitute with 0.1 ml distilled water. Add 0.09% Sodium Azide for long term storage.
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.