Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL12/SDF-1 alpha Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB220507 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB220507 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB220507 Supplier Novus Biologicals Supplier No. NBP229480H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CXCL12/SDF-1 alpha Polyclonal antibody specifically detects CXCL12/SDF-1 alpha in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen CXCL12/SDF-1 alpha
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CXCL12 chemokine (C-X-C motif) ligand 12, IRH, PBSF, SCYB12, SDF1, SDF1A, SDF1B, TLSF, TPAR1
Host Species Rabbit
Immunogen Antisera to human SDF 1 alpha were raised by repeated immunisations of rabbits with highly purified antigen. Purified IgG prepared by affinity chromatography.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 6387
Target Species Human, Rat, Rat, Hamster, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.