Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL12/SDF-1 Antibody (79018) [HRP], Novus Biologicals™
SDP

Catalog No. NB372156 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372156 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372156 Supplier Novus Biologicals Supplier No. FAB350H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCL12/SDF-1 Monoclonal antibody specifically detects CXCL12/SDF-1 in Human, Mouse samples. It is validated for Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF, Dual RNAscope ISH-IHC

Specifications

Antigen CXCL12/SDF-1
Applications Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF, In Situ Hybridization (ISH)
Classification Monoclonal
Clone 79018
Conjugate HRP
Dilution Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Dual RNAscope ISH-IHC
Formulation PBS
Gene Alias chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL12/SDF-1 alpha , Lys22-Lys89, Accession # Q6ICW0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6387
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.