Learn More
CXCL12/SDF-1 Antibody (MM0211-9N26) - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | CXCL12/SDF-1 |
| Applications | CyTOF, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunohistochemistry (Frozen), Flow Cytometry |
| Classification | Monoclonal |
| Clone | MM0211-9N26 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:20-200, Immunohistochemistry 1:50-100, Immunohistochemistry-Paraffin 1:50-100, Immunohistochemistry-Frozen, CyTOF-ready |
| Formulation | A 0.2 um filtered solution in PBS with No Preservative |
| Gene Accession No. | Q6ICW0 |
| Gene Alias | chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.