Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL12/SDF-1 Antibody (MM0211-9N26), PerCP, Novus Biologicals™
SDP

Catalog No. NBP212221P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212221P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212221P Supplier Novus Biologicals Supplier No. NBP212221PCP

Mouse Monoclonal Antibody

CXCL12/SDF-1 Monoclonal specifically detects CXCL12/SDF-1 in Human, Rat, Sheep samples. It is validated for Flow Cytometry.

Specifications

Antigen CXCL12/SDF-1
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0211-9N26
Conjugate PerCP
Dilution Flow Cytometry
Gene Accession No. Q6ICW0
Gene Alias chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1
Gene Symbols CXCL12
Host Species Mouse
Immunogen Human recombinant CXCL12/SDF-1
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6387
Test Specificity Detects human CXCL12 (both 1-alpha and 1 beta)
Target Species Human, Rat, Sheep
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.