Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL12/SDF-1 beta Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB328239 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB328239 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB328239 Supplier Novus Biologicals Supplier No. NBP305873JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CXCL12/SDF-1 beta Polyclonal antibody specifically detects CXCL12/SDF-1 beta in Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen CXCL12/SDF-1 beta
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias hSDF-1beta, SDF 1beta, SDF1B
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse CXCL12/SDF-1 beta (Uniprot#: P40224-1; Lys22-Lys89)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 6387
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.