Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL13/BLC/BCA-1 Antibody (53602), FITC, Novus Biologicals™
SDP

Catalog No. NB295654 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295654 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295654 Supplier Novus Biologicals Supplier No. FAB8012F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCL13/BLC/BCA-1 Monoclonal specifically detects CXCL13/BLC/BCA-1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunohistochemistry, Neutralization.

Specifications

Antigen CXCL13/BLC/BCA-1
Applications Flow Cytometry (Intracellular Staining), Immunohistochemistry, Neutralization
Classification Monoclonal
Clone 53602
Conjugate FITC
Dilution Immunohistochemistry, Neutralization, Intracellular Staining by Flow Cytometry
Formulation PBS
Gene Alias ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant)
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL13/BLC/BCA-1, Val23-Arg94, Accession # O43927
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10563
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.