Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL15/Lungkine Antibody (RM0207-16M7), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB360938 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360938 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360938 Supplier Novus Biologicals Supplier No. NBP212416MFV500
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CXCL15/Lungkine Monoclonal antibody specifically detects CXCL15/Lungkine in Mouse samples. It is validated for Western Blot

Specifications

Antigen CXCL15/Lungkine
Applications Western Blot
Classification Monoclonal
Clone RM0207-16M7
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Il8, IL-8, lungkine, Scyb15, weche
Host Species Rat
Immunogen Mouse recombinant CXCL15/Lungkine
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20309
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.