Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL16 Antibody (256213), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB345611 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345611 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345611 Supplier Novus Biologicals Supplier No. FAB976MFV610
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CXCL16 Monoclonal specifically detects CXCL16 in Human samples. It is validated for CyTOF, Flow Cytometry, Neutralization, Western Blot.

Specifications

Antigen CXCL16
Applications CyTOF, Flow Cytometry, Neutralization, Western Blot
Classification Monoclonal
Clone 256213
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, Neutralization, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16
Host Species Rat
Immunogen E. coli-derived recombinant human CXCL16, Asn30-Pro118, Accession # Q9H2A7.4
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 58191
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.