Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL16 Antibody (RM0208-15Q2) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP212226 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212226 0.1 mg
NB019115 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP212226 Supplier Novus Biologicals Supplier No. NBP212226

Rat Monoclonal Antibody

CXCL16 Monoclonal specifically detects CXCL16 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.

Specifications

Antigen CXCL16
Applications Western Blot, Immunohistochemistry (Paraffin), Blocking Assay
Classification Monoclonal
Clone RM0208-15Q2
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:250-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Accession No. Q8BSU2
Gene Alias chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16
Gene Symbols CXCL16
Host Species Rat
Immunogen Mouse recombinant CXCL16
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58191
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.