Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL16 Antibody (RM0208-15Q2), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB361212 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361212 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361212 Supplier Novus Biologicals Supplier No. NBP212226MFV450

Rat Monoclonal Antibody

CXCL16 Monoclonal antibody specifically detects CXCL16 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization

Specifications

Antigen CXCL16
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
Classification Monoclonal
Clone RM0208-15Q2
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16
Host Species Rat
Immunogen Mouse recombinant CXCL16
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 58191
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.