Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech CXCL16 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. p-7245245
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-8615 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-8615 Supplier PeproTech Supplier No. 500P200BT500UG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: NEGSVTGSCY CGKRISSDSP PSVQFMNRLR KHLRAYHRCL YYTRFQLLSW SVCGGNKDPW VQELMSCLDL KECGHAYSGI VAHQKHLLP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human CXCL16. Anti-Human CXCL16-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hCXCL16 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human CXCL16 (500-P200) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hCXCL16. Western Blot: To detect Human CXCL16 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human CXCL16 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P200BT-1MG will be provided as 2 x 500 μg

CXCL16 is a member of the CXC chemokine family and signals through the CXCR6 receptor. CXCL16 may play a role in attracting lymphocyte subsets during inflammation and may facilitate certain immune responses. The chemokine domain of CXCL16 contains six cysteine residues, including the four highly conserved cysteine residues characteristic of CXC chemokines. The CXCL16 gene codes for a 273 amino acid polypeptide, which includes a 29 amino acid cytoplasmic domain and transmembrane sequence containing approximately 20 amino acids. The extracellular portion of CXCL16 contains a chemokines domain and an extended C-terminal mucin-like stalk sequence. The extracellular domain contains 89 amino acid residues (86 a.a. residues for the murine homolog).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL16
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene Cxcl16
Gene Accession No. Q9H2A7
Gene Alias 0910001K24Rik; AV290116; b2b498Clo; BB024863; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 16; CXC; Cxc chemokine ligand 16; CXC motif chemokine; C-X-C motif chemokine 16; C-X-C motif chemokine ligand 16; CXCL; Cxcl16; CXCL16v1; CXCL16v2; CXCLG16; Mutant line 498; scavenger receptor for phosphatidylserine and oxidized low density lipoprotein; SCYB16; Small-inducible cytokine B16; Srpsox; SR-PSOX; SR-PSOX/CXCL16; Transmembrane chemokine CXCL16; UNQ2759/PRO6714; zinc finger, MYND-type containing 15; Zmynd15
Gene Symbols Cxcl16
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human CXCL16.
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58191
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.