Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL17/VCC-1 Antibody (422208), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB303669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303669 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303669 Supplier Novus Biologicals Supplier No. FAB4207G

Mouse Monoclonal Antibody

CXCL17/VCC-1 Monoclonal antibody specifically detects CXCL17/VCC-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen CXCL17/VCC-1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 422208
Conjugate Alexa Fluor 488
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Chemokine (C-X-C Motif) Ligand 17, C-X-C Motif Chemokine 17, Dcip1, Dendritic Cell And Monocyte Chemokine-Like Protein, DMC, UNQ473, VCC1, VCC-1, VEGF Coregulated Chemokine 1, VEGF Co-Regulated Chemokine 1
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL17/VCC-1, Leu24-Leu119, Accession # Q6UXB2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 284340
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.