Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL2/GRO beta/MIP-2/CINC-3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB325692 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325692 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325692 Supplier Novus Biologicals Supplier No. NBP299401MFV450

Rabbit Polyclonal Antibody

CXCL2/GRO beta/MIP-2/CINC-3 Polyclonal antibody specifically detects CXCL2/GRO beta/MIP-2/CINC-3 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen CXCL2/GRO beta/MIP-2/CINC-3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias chemokine (C-X-C motif) ligand 2, CINC-2a, C-X-C Motif Chemokine 2, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP2, MIP-2a, MIP2A, MIP2-alpha, SCYB2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human CXCL2/GRO beta/MIP-2/CINC-3 (Accession#: NP_002080.1; Ala35-Asn107)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2920
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.