Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL4L1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP17051020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17051020 20 μL
NBP170510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17051020 Supplier Novus Biologicals Supplier No. NBP17051020UL

Rabbit Polyclonal Antibody

CXCL4L1 Polyclonal specifically detects CXCL4L1 in Human samples. It is validated for Western Blot.

Specifications

Antigen CXCL4L1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Alias CXCL4V1, PF4A, PF4-ALT, platelet factor 4 variant, platelet factor 4 variant 1, variant 1 (PF4-like)
Gene Symbols PF4V1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to PF4V1(platelet factor 4 variant 1) The peptide sequence was selected from the middle region of PF4V1. Peptide sequence RPRHITSLEVIKAGPHCPTAQLIATLKNGRKICLDLQALLYKKIIKEHLE.
Molecular Weight of Antigen 11 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5197
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.