Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL5/ENA-78 Antibody (32B3), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB360817 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360817 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360817 Supplier Novus Biologicals Supplier No. NBP222026MFV450

Rat Monoclonal Antibody

CXCL5/ENA-78 Monoclonal antibody specifically detects CXCL5/ENA-78 in Mouse samples. It is validated for Western Blot, ELISA, Neutralization

Specifications

Antigen CXCL5/ENA-78
Applications Western Blot, ELISA, Neutralization
Classification Monoclonal
Clone 32B3
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78
Host Species Rat
Immunogen Mouse recombinant CXCL5/ENA-78
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6374
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.