Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL5/ENA-78 Antibody (33170), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB376311 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376311 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376311 Supplier Novus Biologicals Supplier No. FAB654W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCL5/ENA-78 Monoclonal antibody specifically detects CXCL5/ENA-78 in Human samples. It is validated for ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen CXCL5/ENA-78
Applications ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 33170
Conjugate DyLight 650
Dilution ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL5/ENA-78, Ala37-Asn114, Accession # P42830
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6374
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.