Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL5/ENA-78 Antibody (MM0216-10L19), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB212231405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB212231405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB212231405 Supplier Novus Biologicals Supplier No. NBP212231AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CXCL5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0216-10L19
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias chemokine (C-X-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), epithelial-derived neutrophil activating protein 78, epithelial-derived neutrophil-activating peptide 78, Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78, small inducible cytokine B5, small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derivedneutrophil-activating peptide 78), Small-inducible cytokine B5
Gene Symbols CXCL5
Host Species Mouse
Immunogen Human recombinant CXCL5/ENA-78
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6374
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.