Learn More
CXCL5/ENA-78 Antibody (MM0216-10L19) - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | CXCL5 |
| Applications | Immunohistochemistry, Blocking Assay, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | MM0216-10L19 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Blocking/Neutralizing 50-200 molar excess |
| Formulation | A 0.2 um filtered solution in PBS with No Preservative |
| Gene Accession No. | P42830 |
| Gene Alias | chemokine (C-X-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), epithelial-derived neutrophil activating protein 78, epithelial-derived neutrophil-activating peptide 78, Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78, small inducible cytokine B5, small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derivedneutrophil-activating peptide 78), Small-inducible cytokine B5 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.