Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL6/GCP-2 Antibody (MM0217-10L25), PerCP, Novus Biologicals™
SDP

Catalog No. NB361178 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361178 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361178 Supplier Novus Biologicals Supplier No. NBP212232PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCL6/GCP-2 Monoclonal antibody specifically detects CXCL6/GCP-2 in Human samples. It is validated for Western Blot, Neutralization

Specifications

Antigen CXCL6/GCP-2
Applications Western Blot, Neutralization
Classification Monoclonal
Clone MM0217-10L25
Conjugate PerCP
Dilution Western Blot, Block/Neutralize
Formulation PBS
Gene Alias chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2), Chemokine alpha 3, CKA-3Granulocyte chemotactic protein 2, GCP2member 6 (granulocytechemotactic protein 2), GCP-2Small-inducible cytokine B6, Small inducible cytokine subfamily B (Cys-X-Cys), member b
Host Species Mouse
Immunogen Human recombinant CXCL6/GCP-2
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6372
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.