Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL9/MIG Antibody (050), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB067693 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB067693 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB067693 Supplier Novus Biologicals Supplier No. NBP289697AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CXCL9/MIG Monoclonal specifically detects CXCL9/MIG in Human samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen CXCL9/MIG
Applications ELISA
Classification Monoclonal
Clone 50
Conjugate Alexa Fluor 594
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CXCL9/MIG (Q07325; Thr23-Thr125).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4283
Test Specificity Has cross-reactivity in ELISA with: Mouse CXCL9 Cynomolgus CXCL9 No cross-reactivity in ELISA with: Human CXCL10
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.