Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL9/MIG Antibody (277), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB333192 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333192 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333192 Supplier Novus Biologicals Supplier No. NBP290453APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CXCL9/MIG Monoclonal antibody specifically detects CXCL9/MIG in Mouse samples. It is validated for ELISA

Specifications

Antigen CXCL9/MIG
Applications ELISA
Classification Monoclonal
Clone 277
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse CXCL9/MIG (Uniprot#: P18340, Thr22-Thr126)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4283
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.