Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR1/IL-8RA Antibody (42709) [Alexa Fluor™ 532], Novus Biologicals™
SDP

Catalog No. NB372616 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372616 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372616 Supplier Novus Biologicals Supplier No. FAB3302X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCR1/IL-8RA Monoclonal specifically detects CXCR1/IL-8RA in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen CXCR1/IL-8RA
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 42709
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CXCR1/IL-8 RA, Met1-Leu350, Accession # AAA59159
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3577
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.