Learn More
Description
Specifications
Specifications
| Antigen | CXCR1/IL-8RA |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 8F1-1-4 |
| Conjugate | DyLight 755 |
| Dilution | Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A |
| Gene Symbols | CXCR1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
